Merck
  • Identification of an activation site in Bak and mitochondrial Bax triggered by antibodies.
Identification of an activation site in Bak and mitochondrial Bax triggered by antibodies.
Nature communications (2016-05-25)
Sweta Iyer, Khatira Anwari, Amber E Alsop, Wai Shan Yuen, David C S Huang, John Carroll, Nicholas A Smith, Brian J Smith, Grant Dewson, Ruth M Kluck
ABSTRACT
During apoptosis, Bak and Bax are activated by BH3-only proteins binding to the α2-α5 hydrophobic groove; Bax is also activated via a rear pocket. Here we report that antibodies can directly activate Bak and mitochondrial Bax by binding to the α1-α2 loop. A monoclonal antibody (clone 7D10) binds close to α1 in non-activated Bak to induce conformational change, oligomerization, and cytochrome c release. Anti-FLAG antibodies also activate Bak containing a FLAG epitope close to α1. An antibody (clone 3C10) to the Bax α1-α2 loop activates mitochondrial Bax, but blocks translocation of cytosolic Bax. Tethers within Bak show that 7D10 binding directly extricates α1; a structural model of the 7D10 Fab bound to Bak reveals the formation of a cavity under α1. Our identification of the α1-α2 loop as an activation site in Bak paves the way to develop intrabodies or small molecules that directly and selectively regulate these proteins.
Sigma-Aldrich
Anti-Bak antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution