Merck
Acceptor-free alcohol dehydrogenation by recyclable ruthenium catalyst.
Organic letters (2006-06-02)
Won-Hee Kim, In Soo Park, Jaiwook Park
ABSTRACT
An efficient oxidant-free oxidation for a wide range of alcohols was achieved by a recyclable ruthenium catalyst. The catalyst was prepared from readily available reagents by a one-pot synthesis through nanoparticle generation and gelation. [structure: see text]
Sigma-Aldrich
Ruthenium(III) chloride hydrate, 99.98% trace metals basis