Merck
  • Up-regulation of TDAG51 is a dependent factor of LPS-induced RAW264.7 macrophages proliferation and cell cycle progression.
Up-regulation of TDAG51 is a dependent factor of LPS-induced RAW264.7 macrophages proliferation and cell cycle progression.
Immunopharmacology and immunotoxicology (2016-02-14)
Han-Wei Jiao, Xiao-Xiao Jia, Tian-Jing Zhao, Hui Rong, Jia-Ning Zhang, Ying Cheng, Hua-Pei Zhu, Kai-Lian Xu, Shi-Yu Guo, Qiao-Yun Shi, Hui Zhang, Feng-Yang Wang, Chuang-Fu Chen, Li Du
ABSTRACT
As a component of the outer membrane in Gram-negative bacteria, lipopolysaccharide (LPS)-induced proliferation and cell cycle progression of monocytes/macrophages. It has been suggested that the proapoptotic T-cell death-associated gene 51 (TDAG51) might be associated with cell proliferation and cell cycle progression; however, its role in the interaction between LPS and macrophages remains unclear. We attempted to elucidate the role(s) of TDAG51 played in the interaction between LPS and macrophages. We investigated TDAG51 expression in RAW264.7 cells stimulated with LPS and examined the effects of RNA interference-mediated TDAG51 down-regulation. We used CCK-8 assay and flow cytometry analysis to evaluate the interaction between TDAG51 and LPS-induced proliferation and cell cycle progression in RAW264.7 cells. Our findings indicate that TDAG51 is up-regulated in LPS-stimulated RAW264.7 cells, the TDAG51 siRNA effectively reduced TDAG51 protein up-regulation following LPS stimulation in RAW264.7 cells, the significant changes of the proliferation and cell cycle progression of RAW264.7 cells in TDAG51 Knockdown RAW264.7 cells treated with LPS were observed. These findings suggested that TDAG51 up-regulation is a dependent event during LPS-mediated proliferation and cell cycle progression, and which increase our understanding of the interaction mechanism between LPS and macrophages.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human PHLDA1