Merck
Silylative Dieckmann-like cyclizations of ester-imides (and diesters).
Organic letters (2006-11-03)
Thomas R Hoye, Vadims Dvornikovs, Elena Sizova
ABSTRACT
[Structure: see text] Trialkylsilyl triflates effect cyclization of ester-imides such as 2 to produce adducts such as 4a. Trapping of the in situ generated, nucleophilic ketene acetal (cf. 5a) is a key aspect of the transformation. A range of substrates amenable to this operationally simple reaction is reported. In many instances the levels of diastereoselectivity are very high. Mechanistic points are inferred from spectroscopic observations.
Sigma-Aldrich
Trimethylsilyl trifluoromethanesulfonate, 99%