Merck
[3,3]-rearrangements of phosphonium ylides.
Journal of the American Chemical Society (2006-04-06)
Marcelle L Ferguson, Todd D Senecal, Todd M Groendyke, Anna K Mapp
ABSTRACT
Allylic phosphonium ylides are readily generated by the combination of an allylic alcohol, a carbene, and a chlorophosphite. Here we demonstrate that these intermediates undergo a thermal [3,3]-rearrangement to provide single isomers of homoallylic phosphonates in good to excellent yields. This new reaction manifold for phosphorus ylides is tolerant of a range of substitution patterns on the reactants and provides access to structurally complex intermediates for the synthesis of enzyme inhibitors, aminophosphonic acids, and natural products.
Sigma-Aldrich
Diisopropyl azodicarboxylate, 98%