Merck
  • Identification of a functional variant in the MICA promoter which regulates MICA expression and increases HCV-related hepatocellular carcinoma risk.
Identification of a functional variant in the MICA promoter which regulates MICA expression and increases HCV-related hepatocellular carcinoma risk.
PloS one (2013-04-18)
Paulisally Hau Yi Lo, Yuji Urabe, Vinod Kumar, Chizu Tanikawa, Kazuhiko Koike, Naoya Kato, Daiki Miki, Kazuaki Chayama, Michiaki Kubo, Yusuke Nakamura, Koichi Matsuda
ABSTRACT
Hepatitis C virus (HCV) infection is the major cause of hepatocellular carcinoma (HCC) in Japan. We previously identified the association of SNP rs2596542 in the 5' flanking region of the MHC class I polypeptide-related sequence A (MICA) gene with the risk of HCV-induced HCC. In the current study, we performed detailed functional analysis of 12 candidate SNPs in the promoter region and found that a SNP rs2596538 located at 2.8 kb upstream of the MICA gene affected the binding of a nuclear protein(s) to the genomic segment including this SNP. By electrophoretic mobility shift assay (EMSA) and chromatin immunoprecipitation (ChIP) assay, we identified that transcription factor Specificity Protein 1 (SP1) can bind to the protective G allele, but not to the risk A allele. In addition, reporter construct containing the G allele was found to exhibit higher transcriptional activity than that containing the A allele. Moreover, SNP rs2596538 showed stronger association with HCV-induced HCC (P = 1.82 × 10(-5) and OR = 1.34) than the previously identified SNP rs2596542. We also found significantly higher serum level of soluble MICA (sMICA) in HCV-induced HCC patients carrying the G allele than those carrying the A allele (P = 0.00616). In summary, we have identified a functional SNP that is associated with the expression of MICA and the risk for HCV-induced HCC.