- Colonic MUC2 mucin regulates the expression and antimicrobial activity of β-defensin 2.
Colonic MUC2 mucin regulates the expression and antimicrobial activity of β-defensin 2.
In this study we identified mechanisms at the colonic mucosa by which MUC2 mucin regulated the production of β-defensin in a proinflammatory milieu but functionally protected susceptible bacteria from its antimicrobial effects. The regulator role of MUC2 on production of β-defensin 2 in combination with the proinflammatory cytokine interleukin-1β (IL-1β) was confirmed using purified human colonic MUC2 mucin and colonic goblet cells short hairpin RNA (shRNA) silenced for MUC2. In vivo, Muc2(-/-) mice showed impaired β-defensin mRNA expression and peptide localization in the colon as compared with Muc2(+/-) and Muc2(+/+) littermates. Importantly, purified MUC2 mucin abrogated the antimicrobial activity of β-defensin 2 against nonpathogenic and enteropathogenic Escherichia coli. Sodium metaperiodate oxidation of MUC2 removed the capacity of MUC2 to stimulate β-defensin production and MUC2's inhibition of defensin antimicrobial activity. This study highlights that a defective MUC2 mucin barrier, typical in inflammatory bowel diseases, may lead to deficient stimulation of β-defensin 2 and an unbalanced microbiota that favor the growth of β-defensin-resistant microbes such as Clostridium difficile.