Merck
  • NuSAP modulates the dynamics of kinetochore microtubules by attenuating MCAK depolymerisation activity.
NuSAP modulates the dynamics of kinetochore microtubules by attenuating MCAK depolymerisation activity.
Scientific reports (2016-01-07)
Chenyu Li, Yajun Zhang, Qiaoyun Yang, Fan Ye, Stella Ying Sun, Ee Sin Chen, Yih-Cherng Liou
ABSTRACT
Nucleolar and spindle-associated protein (NuSAP) is a microtubule-associated protein that functions as a microtubule stabiliser. Depletion of NuSAP leads to severe mitotic defects, however the mechanism by which NuSAP regulates mitosis remains elusive. In this study, we identify the microtubule depolymeriser, mitotic centromere-associated kinesin (MCAK), as a novel binding partner of NuSAP. We show that NuSAP regulates the dynamics and depolymerisation activity of MCAK. Phosphorylation of MCAK by Aurora B kinase, a component of the chromosomal passenger complex, significantly enhances the interaction of NuSAP with MCAK and modulates the effects of NuSAP on the depolymerisation activity of MCAK. Our results reveal an underlying mechanism by which NuSAP controls kinetochore microtubule dynamics spatially and temporally by modulating the depolymerisation function of MCAK in an Aurora B kinase-dependent manner. Hence, this study provides new insights into the function of NuSAP in spindle formation during mitosis.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
MISSION® esiRNA, targeting human KIF2C