Merck
  • Genetically engineered flavonol enriched tomato fruit modulates chondrogenesis to increase bone length in growing animals.
Genetically engineered flavonol enriched tomato fruit modulates chondrogenesis to increase bone length in growing animals.
Scientific reports (2016-02-27)
Dharmendra Choudhary, Ashutosh Pandey, Sulekha Adhikary, Naseer Ahmad, Chitra Bhatia, Sweta Bhambhani, Prabodh Kumar Trivedi, Ritu Trivedi
ABSTRACT
Externally visible body and longitudinal bone growth is a result of proliferation of chondrocytes. In growth disorder, there is delay in the age associated increase in height. The present study evaluates the effect of extract from transgenic tomato fruit expressing AtMYB12 transcription factor on bone health including longitudinal growth. Constitutive expression of AtMYB12 in tomato led to a significantly enhanced biosynthesis of flavonoids in general and the flavonol biosynthesis in particular. Pre-pubertal ovary intact BALB/c mice received daily oral administration of vehicle and ethanolic extract of wild type (WT-TOM) and transgenic AtMYB12-tomato (MYB12-TOM) fruits for six weeks. Animal fed with MYB12-TOM showed no inflammation in hepatic tissues and normal sinusoidal Kupffer cell morphology. MYB12-TOM extract significantly increased tibial and femoral growth and subsequently improved the bone length as compared to vehicle and WT-TOM. Histomorphometry exhibited significantly wider distal femoral and proximal tibial growth plate, increased number and size of hypertrophic chondrocytes in MYB12-TOM which corroborated with micro-CT and expression of BMP-2 and COL-10, marker genes for hypertrophic cells. We conclude that metabolic reprogramming of tomato by AtMYB12 has the potential to improve longitudinal bone growth thus helping in achievement of greater peak bone mass during adolescence.
Sigma-Aldrich
Hematoxylin solution according to Mayer