Merck
  • Photodynamic therapy of tumours with hexadecafluoro zinc phthalocynine formulated in PEG-coated poly(lactic acid) nanoparticles.
Photodynamic therapy of tumours with hexadecafluoro zinc phthalocynine formulated in PEG-coated poly(lactic acid) nanoparticles.
International journal of cancer (1996-06-11)
E Allémann, J Rousseau, N Brasseur, S V Kudrevich, K Lewis, J E van Lier
ABSTRACT
Hexadecafluoro zinc phthalocyanine (ZnPcF16), a second-generation sensitizer for the photodynamic therapy (PDT) of cancer, was formulated in polyethylene-glycol-coated poly(lactic acid) nanoparticles (PEG-coated PLA-NP) and tested in EMT-6 tumour-bearing mice for its photodynamic activity. The tumour response was compared to that induced by the same dye formulated as a Cremophor EL (CRM) emulsion. Formulation in the biodegradable NP improved PDT response of the tumour while providing prolonged tumour sensitivity towards PDT.
Sigma-Aldrich
Zinc 1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-hexadecafluoro-29H,31H-phthalocyanine, Dye content 90 %