Merck
  • A rapid and convenient method for fluorescence analysis of in vitro cultivated metacestode vesicles from Echinococcus multilocularis.
A rapid and convenient method for fluorescence analysis of in vitro cultivated metacestode vesicles from Echinococcus multilocularis.
PloS one (2015-02-24)
Zhe Cheng, Fan Liu, Shan Zhu, Huimin Tian, Liang Wang, Yanhai Wang
ABSTRACT
We here describe a convenient method for preparation, fixation and fluorescence analysis of in vitro cultivated metacestode vesicles from E. multilocularis. Parasite materials could be prepared in one hour, did not need to be sectioned, and were subsequently utilized for further whole-mount staining assays directly. Using these preparations, in combination with conventional fluorescence staining techniques, we could detect the expression and subcellular localization of a specific protein and identify in situ proliferative or apoptotic cells in the germinal layer of metacestode vesicles. Based on this approach, future molecular and cellular analysis of Echinococcus metacestode vesicles in the in vitro system will be greatly facilitated.
Product Description
Roche
In Situ Cell Death Detection Kit, AP, sufficient for ≤50 tests, kit of 1 (3 components), suitable for detection