Merck
  • Impaired photoreceptor protein transport and synaptic transmission in a mouse model of Bardet-Biedl syndrome.
Impaired photoreceptor protein transport and synaptic transmission in a mouse model of Bardet-Biedl syndrome.
Vision research (2007-11-21)
Muhammad M Abd-El-Barr, Kristen Sykoudis, Sara Andrabi, Erica R Eichers, Mark E Pennesi, Perciliz L Tan, John H Wilson, Nicholas Katsanis, James R Lupski, Samuel M Wu
ABSTRACT
Bardet-Biedl syndrome (BBS) is an oligogenic syndrome whose manifestations include retinal degeneration, renal abnormalities, obesity and polydactylia. Evidence suggests that the main etiopathophysiology of this syndrome is impaired intraflagellar transport (IFT). In this study, we study the Bbs4-null mouse and investigate photoreceptor structure and function after loss of this gene. We find that Bbs4-null mice have defects in the transport of phototransduction proteins from the inner segments to the outer segments, before signs of cell death. Additionally, we show defects in synaptic transmission from the photoreceptors to secondary neurons of the visual system, demonstrating multiple functions for BBS4 in photoreceptors.
Product Description
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection
Pricing and availability is not currently available.