Merck
  • Photoclick Hydrogels Prepared from Functionalized Cyclodextrin and Poly(ethylene glycol) for Drug Delivery and in Situ Cell Encapsulation.
Photoclick Hydrogels Prepared from Functionalized Cyclodextrin and Poly(ethylene glycol) for Drug Delivery and in Situ Cell Encapsulation.
Biomacromolecules (2015-05-23)
Han Shih, Chien-Chi Lin
ABSTRACT
Polymers or hydrogels containing modified cyclodextrin (CD) are highly useful in drug delivery applications, as CD is a cytocompatible amphiphilic molecule that can complex with a variety of hydrophobic drugs. Here, we designed modular photoclick thiol-ene hydrogels from derivatives of βCD and poly(ethylene glycol) (PEG), including βCD-allylether (βCD-AE), βCD-thiol (βCD-SH), PEG-thiol (PEGSH), and PEG-norbornene (PEGNB). Two types of CD-PEG hybrid hydrogels were prepared using radical-mediated thiol-ene photoclick reactions. Specifically, thiol-allylether hydrogels were formed by reacting multiarm PEGSH and βCD-AE, and thiol-norbornene hydrogels were formed by cross-linking βCD-SH and multiarm PEGNB. We characterized the properties of these two types of thiol-ene hydrogels, including gelation kinetics, gel fractions, hydrolytic stability, and cytocompatibility. Compared with thiol-allylether hydrogels, thiol-norbornene photoclick reaction formed hydrogels with faster gelation kinetics at equivalent macromer contents. Using curcumin, an anti-inflammatory and anticancer hydrophobic molecule, we demonstrated that CD-cross-linked PEG-based hydrogels, when compared with pure PEG-based hydrogels, afforded higher drug loading efficiency and prolonged delivery in vitro. Cytocompatibility of these CD-cross-linked hydrogels were evaluated by in situ encapsulation of radical sensitive pancreatic MIN6 β-cells. All formulations and cross-linking conditions tested were cytocompatible for cell encapsulation. Furthermore, hydrogels cross-linked by βCD-SH showed enhanced cell proliferation and insulin secretion as compared to gels cross-linked by either dithiothreitol (DTT) or βCD-AE, suggesting the profound impact of both macromer compositions and gelation chemistry on cell fate in chemically cross-linked hydrogels.
Sigma-Aldrich
Allyl bromide, reagent grade, 97%, contains ≤1000 ppm propylene oxide as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydride, dry, 90%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Pricing and availability is not currently available.
Sigma-Aldrich
Allyl bromide, ReagentPlus®, 99%, contains ≤1000 ppm propylene oxide as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
DCC, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Pyridine, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
DCC, 1.0 M in methylene chloride
Pricing and availability is not currently available.
Sigma-Aldrich
Hydriodic acid, 57 wt. %, distilled, 99.999% trace metals basis
Pricing and availability is not currently available.