Merck
CCR5-CCL Axis in PDL during Orthodontic Biophysical Force Application.
Journal of dental research (2015-09-12)
S Y Lee, H I Yoo, S H Kim
ABSTRACT
Tooth movement by application of orthodontic biophysical force primarily reflects the role of soluble molecules released from the periodontal ligament (PDL). Thus far, many factors have been reported to be involved in orthodontic tooth movement (OTM), but key molecules that orchestrate responses of periodontal tissues to biophysical force are still enigmatic. In this in vivo study, in which the upper first molars in rats were moved, differential display-polymerase chain reaction revealed that CC chemokine receptor 5 (CCR5) level was differentially increased during OTM. Strong immunoreactivity for CCR5 was found in the PDL undergoing force application. Moreover, the in vitro compression or tension force application to primary cultured human PDL cells increased the expression of CCR5 and CCR5 ligands. In vitro tension force on human PDL cells did not induce RANKL, an osteoclastogenesis-inducing factor, but did induce the upregulation of IL12, an osteoclast inhibitory factor, and osteoblast differentiation factors, including Runx2, which was attenuated under tension by CCR5 gene silencing whereas augmented with CCR5 ligands. In contrast, in vitro compression force did not induce the expression of osteoprotegerin, a decoy receptor for RANKL and Runx2, but did induce the upregulation of RANKL, which was attenuated under compression by CCR5 gene silencing. These results suggest that the CCR5-CCR5 ligands axis in PDL cells may play a crucial role in the remodeling of periodontal tissues and can be a therapeutic target for achieving efficient OTM.
Sigma-Aldrich
Diethylene glycol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Diethylene glycol, puriss. p.a., ≥99.0% (GC), colorless
Sigma-Aldrich
Diethylene glycol, ReagentPlus®, 99%
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ccr5
Sigma-Aldrich
MISSION® esiRNA, targeting human CCR5