Merck
  • Extensively stereodiversified scaffolds for use in diversity-oriented library synthesis.
Extensively stereodiversified scaffolds for use in diversity-oriented library synthesis.
Organic letters (2003-02-28)
Tiffany Malinky Gierasch, Zhangjie Shi, Gregory L Verdine
ABSTRACT
The syntheses of stereodiverse libraries of 12 and 19 are reported, where each asterisk represents an independently varied stereocenter. These scaffolds provide additional templates for investigations of geometric diversity in library syntheses. Libraries of these N-Fmoc-amino acids were further functionalized by incorporation into a peptide sequence, demonstrating the utility of 12 and 19 as building blocks for diversity oriented synthesis.