Merck
  • Effects of 8-methoxypsoralen on the metabolism of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone in mice.
Effects of 8-methoxypsoralen on the metabolism of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone in mice.
Drug metabolism and pharmacokinetics (2015-07-27)
Mingfei Feng, Hongtu Zhang, Baojun Cao, Shuaidong Liu, Jian Mao, Qidong Zhang
ABSTRACT
8-Methoxypsoralen (8-MOP) is a well established drug in the treatment of various skin diseases. Pretreatment of mice with 8-MOP before administration of 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) significantly reduced the incidence of NNK-induced tumor. The present study was designed to evaluate the in vivo effects of 8-MOP on the bioactivation of NNK in mice. Decrease in the α-hydroxylation of NNK in mouse blood and tissues was observed as the most pronounced effect of 8-MOP. The catalytic property of cytochrome P450 2A5 (CYP2A5) enzyme in mice was determined by the coumarin 7-hydroxylation reaction, suggesting that 8-MOP produced remarkable inhibition on CYP2A5 in female C57BL/6 mice. These results implied that 8-MOP could prevent NNK-induced mutagenesis and tumorigenesis in mice through the inhibition of NNK α-hydroxylation, which may be achieved through the effect of 8-MOP on the bioactivities of CYP2A5.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Coumarin, ≥99% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Umbelliferone, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N