Merck
  • Integrated metabolomics and transcriptomics reveal enhanced specialized metabolism in Medicago truncatula root border cells.
Integrated metabolomics and transcriptomics reveal enhanced specialized metabolism in Medicago truncatula root border cells.
Plant physiology (2015-02-11)
Bonnie S Watson, Mohamed F Bedair, Ewa Urbanczyk-Wochniak, David V Huhman, Dong Sik Yang, Stacy N Allen, Wensheng Li, Yuhong Tang, Lloyd W Sumner
ABSTRACT
Integrated metabolomics and transcriptomics of Medicago truncatula seedling border cells and root tips revealed substantial metabolic differences between these distinct and spatially segregated root regions. Large differential increases in oxylipin-pathway lipoxygenases and auxin-responsive transcript levels in border cells corresponded to differences in phytohormone and volatile levels compared with adjacent root tips. Morphological examinations of border cells revealed the presence of significant starch deposits that serve as critical energy and carbon reserves, as documented through increased β-amylase transcript levels and associated starch hydrolysis metabolites. A substantial proportion of primary metabolism transcripts were decreased in border cells, while many flavonoid- and triterpenoid-related metabolite and transcript levels were increased dramatically. The cumulative data provide compounding evidence that primary and secondary metabolism are differentially programmed in border cells relative to root tips. Metabolic resources normally destined for growth and development are redirected toward elevated accumulation of specialized metabolites in border cells, resulting in constitutively elevated defense and signaling compounds needed to protect the delicate root cap and signal motile rhizobia required for symbiotic nitrogen fixation. Elevated levels of 7,4'-dihydroxyflavone were further increased in border cells of roots exposed to cotton root rot (Phymatotrichopsis omnivora), and the value of 7,4'-dihydroxyflavone as an antimicrobial compound was demonstrated using in vitro growth inhibition assays. The cumulative and pathway-specific data provide key insights into the metabolic programming of border cells that strongly implicate a more prominent mechanistic role for border cells in plant-microbe signaling, defense, and interactions than envisioned previously.
Sigma-Aldrich
Potassium fluoride, anhydrous, powder, ≥99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Pricing and availability is not currently available.
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium fluoride, ≥99.97% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium fluoride, BioUltra, ≥99.5% (F)
Pricing and availability is not currently available.
Sigma-Aldrich
Divinylbenzene, technical grade, 55%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Pricing and availability is not currently available.
Sigma-Aldrich
Flavone, ≥99.0%
Pricing and availability is not currently available.
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Pricing and availability is not currently available.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Divinylbenzene, technical grade, 80%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Fluorescein diacetate, used as cell viability stain
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Umbelliferone, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Propidium iodide solution
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Allene, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Pricing and availability is not currently available.