- A fluorescent organic cage for picric acid detection.
A fluorescent organic cage for picric acid detection.
Chemical communications (Cambridge, England) (2014-11-06)
Koushik Acharyya, Partha Sarathi Mukherjee
PMID25370367
ABSTRACT
Dynamic covalent imine chemistry has been utilized to synthesize a fluorescent [3+2] self-assembled nanoscopic organic cage. The fluorescent nature of the reduced analogue of the cage was further exploited for the highly selective detection of the explosive picric acid (PA).