Merck
  • Molecular cloning and characterization of a novel human putative transmembrane protein homologous to mouse sideroflexin associated with sideroblastic anemia.
Molecular cloning and characterization of a novel human putative transmembrane protein homologous to mouse sideroflexin associated with sideroblastic anemia.
DNA sequence : the journal of DNA sequencing and mapping (2004-02-06)
Huarui Zheng, Chaoneng Ji, Xianqiong Zou, Maoqing Wu, Zhe Jin, Gang Yin, Jixi Li, Congjing Feng, Haipeng Cheng, Shaohua Gu, Yi Xie, Yumin Mao
ABSTRACT
Sideroflexin1 (Sfxn1), the prototype of a novel family of evolutionarily conserved proteins present in eukaryotes, has been found mutated in mice with siderocytic anemia. It is speculated that this protein facilitates the transport of a component required for iron utilization into mitochondrial. During the large-scale sequencing analysis of a human fetal brain cDNA library, we isolated a cDNA encoding a novel sideroflexin protein (SFXN4), which showed 59% identity and 71% similarity to mouse sideroflexin4. According to the search of the human genome database, SFXN4 gene is mapped to chromosome 10q25-26 and spans more than 24.7kb of the genomic DNA. It is 1428 base pair in length and the putative protein contains 305 amino acids with a conserved predicted five-transmembrane-domains structure. RT-PCR result shows that the SFXN4 gene is expressed in many tissues.