Merck
  • A single gene deletion on 4q28.3: PCDH18--a new candidate gene for intellectual disability?
A single gene deletion on 4q28.3: PCDH18--a new candidate gene for intellectual disability?
European journal of medical genetics (2012-03-28)
Jurate Kasnauskiene, Zivile Ciuladaite, Egle Preiksaitiene, Aušra Matulevičienė, Angelos Alexandrou, George Koumbaris, Carolina Sismani, Ingrida Pepalytė, Philippos C Patsalis, Vaidutis Kučinskas
ABSTRACT
We report a boy with severe developmental delay, seizures, microcephaly, hypoplastic corpus callosum, internal hydrocephalus and dysmorphic features (narrow forehead, round face, deep-set eyes, blue sclerae, large and prominent ears, nose with anteverted nares, thin upper lip, small and wide-spaced teeth, hyperextensibility of the elbows, wrists, and fingers, fingertip pads, broad hallux, sacral dimple), carrying a 1.53 Mb interstitial deletion at 4q28.3. The deletion was detected by Agilent 105K oligo-array genome hybridization and involves the genomic region between 137,417,338 and 138,947,282 base pairs on chromosome 4 (NCBI build 36). The alteration was inherited from a healthy mother and contains a single gene, PCDH18 which encodes a cadherin-related neuronal receptor thought to play a role in the establishment and function of cell-cell connections in the brain. Thus, haploinsufficiency of this gene may contribute to altered brain development and associated malformations. We found that this deletion is a private inherited copy number variation that is associated with specific clinical findings in our patient and propose the PCDH18 gene as a possible candidate gene for intellectual disability.