Merck
  • Candidate tumor suppressor BTG3 maintains genomic stability by promoting Lys63-linked ubiquitination and activation of the checkpoint kinase CHK1.
Candidate tumor suppressor BTG3 maintains genomic stability by promoting Lys63-linked ubiquitination and activation of the checkpoint kinase CHK1.
Proceedings of the National Academy of Sciences of the United States of America (2013-03-28)
Yu-Che Cheng, Tsong-Yu Lin, Sheau-Yann Shieh
ABSTRACT
B-cell translocation gene 3 (BTG3) is a member of the antiproliferative BTG/ Transducer of ErbB2 gene family and is induced by genotoxic stress in a p53- and Checkpoint kinase 1 (CHK1)-dependent manner. Down-regulation of BTG3 has been observed in human cancers, suggesting that it plays an important role in tumor suppression, although the underlying mechanisms are unclear. Here, we report that BTG3 interacts with CHK1, a key effector kinase in the cell cycle checkpoint response, and regulates its phosphorylation and activation. Upon interaction, BTG3 mediates K63-linked ubiquitination of CHK1 at Lys132 through the cullin-RING ligase 4(Cdt2) E3 complex, thus facilitating CHK1 chromatin association. We show that BTG3-depleted cells phenocopy those CHK1-deficient cells, exhibiting increased cell death after replication block and impaired chromosome alignment and segregation. These defects could be corrected by wild-type BTG3 but not by a mutant impaired in CHK1 interaction. We propose that BTG3-dependent CHK1 ubiquitination contributes to its chromatin localization and activation and that a defect in this regulation may increase genome instability and promote tumorigenesis.