Merck
  • Non-overlapping expression of Olig3 and Olig2 in the embryonic neural tube.
Non-overlapping expression of Olig3 and Olig2 in the embryonic neural tube.
Mechanisms of development (2002-04-19)
Hirohide Takebayashi, Toshiaki Ohtsuki, Tsukasa Uchida, Shoko Kawamoto, Kosaku Okubo, Kazuhiro Ikenaka, Masatoshi Takeichi, Osamu Chisaka, Yo-ichi Nabeshima
ABSTRACT
Olig family is a novel sub-family of basic helix-loop-helix transcription factors recently identified. Olig1 and Olig2 were first reported to promote oligodendrocyte differentiation, and later Olig2 was reported to be involved in motoneuron specification as well. Olig3 was isolated as a third member of Olig family, but its precise expression pattern is poorly understood. Here, we describe detailed Olig3 expression analyses in the neural tube of embryonic mice. Olig3 was first detected in the dorsal neural tube from the midbrain/hindbrain boundary to the spinal cord. In E11.5 spinal cord, Olig3 was transiently expressed in the lateral margin of the subventricular zone as three ventral clusters at the level of the p3, p2 and p0 domains, as well as in the dorsal neural tube. Olig3 was co-expressed with Nkx2.2 in the lateral margin of the p3 domain. In forebrain, Olig3 was expressed in the dorsal thalamus while Olig2 was complementarily expressed in the ventral thalamus with an adjacent boundary at E12.5. Olig3 is specifically and transiently expressed in different types of progenitors of embryonic central nervous system and then disappears in the course of development.