Merck
  • Repetitive transcranial magnetic stimulation increases the corticospinal inhibition and the brain-derived neurotrophic factor in chronic myofascial pain syndrome: an explanatory double-blinded, randomized, sham-controlled trial.
Repetitive transcranial magnetic stimulation increases the corticospinal inhibition and the brain-derived neurotrophic factor in chronic myofascial pain syndrome: an explanatory double-blinded, randomized, sham-controlled trial.
The journal of pain (2014-05-29)
Letizzia Dall'Agnol, Liciane Fernandes Medeiros, Iraci L S Torres, Alicia Deitos, Aline Brietzke, Gabriela Laste, Andressa de Souza, Júlia Lima Vieira, Felipe Fregni, Wolnei Caumo
ABSTRACT
Chronic myofascial pain syndrome has been related to defective descending inhibitory systems. Twenty-four females aged 19 to 65 years with chronic myofascial pain syndrome were randomized to receive 10 sessions of repetitive transcranial magnetic stimulation (rTMS) (n = 12) at 10 Hz or a sham intervention (n = 12). We tested if pain (quantitative sensory testing), descending inhibitory systems (conditioned pain modulation [quantitative sensory testing + conditioned pain modulation]), cortical excitability (TMS parameters), and the brain-derived neurotrophic factor (BDNF) would be modified. There was a significant interaction (time vs group) regarding the main outcomes of the pain scores as indexed by the visual analog scale on pain (analysis of variance, P < .01). Post hoc analysis showed that compared with placebo-sham, the treatment reduced daily pain scores by -30.21% (95% confidence interval = -39.23 to -21.20) and analgesic use by -44.56 (-57.46 to -31.67). Compared to sham, rTMS enhanced the corticospinal inhibitory system (41.74% reduction in quantitative sensory testing + conditioned pain modulation, P < .05), reduced the intracortical facilitation in 23.94% (P = .03), increased the motor evoked potential in 52.02% (P = .02), and presented 12.38 ng/mL higher serum BDNF (95% confidence interval = 2.32-22.38). No adverse events were observed. rTMS analgesic effects in chronic myofascial pain syndrome were mediated by top-down regulation mechanisms, enhancing the corticospinal inhibitory system possibly via BDNF secretion modulation. High-frequency rTMS analgesic effects were mediated by top-down regulation mechanisms enhancing the corticospinal inhibitory, and this effect involved an increase in BDNF secretion.