Merck
  • Behavioural profile of exendin-4/naltrexone dose combinations in male rats during tests of palatable food consumption.
Behavioural profile of exendin-4/naltrexone dose combinations in male rats during tests of palatable food consumption.
Psychopharmacology (2014-04-01)
F L Wright, R J Rodgers
ABSTRACT
The glucagon-like peptide 1 receptor (GLP-1R) agonist exendin-4 potently suppresses food intake in animals and humans. However, little is known about the behavioural specificity of this effect either when administered alone or when co-administered with another anorectic agent. The present study characterises the effects of exendin-4, both alone and in combination with naltrexone, on behaviours displayed by male rats during tests with palatable mash. Experiment 1 examined the dose-response effects of exendin-4 (0.025-2.5 μg/kg, IP), while experiment 2 profiled the effects of low-dose combinations of the peptide (0.025 and 0.25 μg/kg) and naltrexone (0.1 mg/kg). In experiment 1, exendin-4 dose dependently suppressed food intake as well as the frequency and rate of eating. However, these effects were accompanied by dose-dependent reductions in all active behaviours and, at 2.5 μg/kg, a large increase in resting and disruption of the behavioural satiety sequence (BSS). In experiment 2, while exendin-4 (0.25 μg/kg) and naltrexone each produced a significant reduction in intake and feeding behaviour (plus an acceleration in the BSS), co-treatment failed to produce stronger effects than those seen in response to either compound alone. Similarities between the behavioural signature of exendin-4 and that previously reported for the emetic agent lithium chloride would suggest that exendin-4 anorexia is related to the aversive effects of the peptide. Furthermore, as low-dose combinations of the peptide with naltrexone failed to produce an additive/synergistic anorectic effect, this particular co-treatment strategy would not appear to have therapeutic significance.
Product Description
Naltrexone hydrochloride, European Pharmacopoeia (EP) Reference Standard