Merck
  • SDF2L1 interacts with the ER-associated degradation machinery and retards the degradation of mutant proinsulin in pancreatic β-cells.
SDF2L1 interacts with the ER-associated degradation machinery and retards the degradation of mutant proinsulin in pancreatic β-cells.
Journal of cell science (2013-02-28)
Akansha Tiwari, Irmgard Schuiki, Liling Zhang, Emma M Allister, Michael B Wheeler, Allen Volchuk
ABSTRACT
Stromal cell-derived factor 2-like 1 (SDF2L1) is an endoplasmic reticulum (ER)-localized protein whose function is undefined. Here we show that SDF2L1 protein levels are increased in response to ER stress-inducing compounds, but not other cell stressors that we tested in insulinoma cell lines. SDF2L1 protein levels were also induced by expression of misfolded proinsulin in insulinoma cells and in islets from diabetic mice. Immunoprecipitation and binding assays demonstrated that SDF2L1 interacts with the ER chaperone GRP78/BiP, the ER-associated degradation (ERAD) machinery and with misfolded proinsulin. Unexpectedly, knockdown of SDF2L1 in INS-1 (insulin 2 C96Y-GFP) cells increased the degradation kinetics of mutant proinsulin, suggesting that SDF2L1 regulates substrate availability for the ERAD system. We suggest that SDF2L1 increases the time that misfolded proteins have to achieve a correctly folded conformation and thus that SDF2L1 can act as a buffer for substrate availability for ERAD in pancreatic β-cells.
Sigma-Aldrich
Anti-Derlin-2 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Protein A antibody produced in mouse, clone SPA-27, ascites fluid