Merck
  • Protonated nitro group as a gas-phase electrophile: experimental and theoretical study of the cyclization of o-nitrodiphenyl ethers, amines, and sulfides.
Protonated nitro group as a gas-phase electrophile: experimental and theoretical study of the cyclization of o-nitrodiphenyl ethers, amines, and sulfides.
Journal of the American Society for Mass Spectrometry (2007-11-03)
Joseph T Moolayil, Mathai George, R Srinivas, Daryl Giblin, Amber Russell, Michael L Gross
ABSTRACT
A novel gas-phase electrophilic cyclization, initiated by the protonation of a nitro group, occurs for 2-nitrophenyl phenyl ether and for the analogous sulfide and amine, leading to heterocyclic intermediates in each case. Subsequently, the cyclic intermediates dissociate via two pathways: (1) unusual step-wise eliminations of two OH radicals to afford heterocyclic cations, [phenoxazine - H](+), [phenothiazine - H](+), and [phenazine + H](+), and (2) expulsion of H(2)O, to yield a heterocyclic ketone, followed by loss of CO. The proposed structures of the gas-phase product ions and reaction mechanisms are supported by chemical substitution, deuterium labeling, accurate mass measurements at high mass resolving power, product-ion mass spectra obtained by tandem mass spectrometry, mass spectra of reference compounds, and molecular orbital calculations. Using a mass spectrometer as a reaction vessel, we demonstrate that, upon protonation, a nitro group becomes an electrophile and participates in cyclization reactions in the gas phase.