Merck
  • N-(3-(phenylcarbamoyl)arylpyrimidine)-5-carboxamides as potent and selective inhibitors of Lck: structure, synthesis and SAR.
N-(3-(phenylcarbamoyl)arylpyrimidine)-5-carboxamides as potent and selective inhibitors of Lck: structure, synthesis and SAR.
Bioorganic & medicinal chemistry letters (2007-12-18)
Holly L Deak, John R Newcomb, Joseph J Nunes, Christina Boucher, Alan C Cheng, Erin F DiMauro, Linda F Epstein, Paul Gallant, Brian L Hodous, Xin Huang, Josie H Lee, Vinod F Patel, Stephen Schneider, Susan M Turci, Xiaotian Zhu
ABSTRACT
N-3-(Phenylcarbamoyl)arylpyrimidine-5-carboxamides are a novel class of selective Lck inhibitors. This series of compounds derives its selectivity from a hydrogen bond with the gatekeeper Thr316 of the enzyme. X-ray co-crystal structural data, structure-activity relationships, and the synthesis of these inhibitors are reported herein.
Sigma-Aldrich
4-Methyl-3-nitrobenzoyl chloride, 99%