- Ritter-type reactions of N-chlorosaccharin: a method for the electrophilic diamination of alkenes.
Ritter-type reactions of N-chlorosaccharin: a method for the electrophilic diamination of alkenes.
Organic letters (2003-08-29)
Kevin I Booker-Milburn, Dominic J Guly, Brian Cox, Panayiotis A Procopiou
PMID12943415
ABSTRACT
[reaction: see text] N-Chlorosaccharin has been shown to undergo electrophilic Ritter-type reactions with alkenes in acetonitrile. The resulting labile beta-chloro sulfonylamidines can be ring-opened and cyclized to imidazolines. Overall this provides a one pot method for the electrophilic diamination of alkenes. Competing aziridine formation as well as allylic chlorination are also observed depending on the nature of the alkene used.
MATERIALS