Merck
Synthesis of racemic 9-methyl-10-hexadecenoic acid.
Chemistry and physics of lipids (1999-04-08)
N M Carballeira, A Sostre, J A Restituyo
ABSTRACT
The marine bacterial fatty acid 9-methyl-10-hexadecenoic acid was conveniently prepared in 6 steps and in a 22% overall yield, starting from commercially available methyl 10-hydroxydecanoate. The naturally occurring fatty acid has the E double bond configuration as confirmed by gas chromatographic co-elution experiments.
Sigma-Aldrich
Methyl 10-hydroxydecanoate, technical grade, ≥90%