Merck
  • Mutations in the SPARC-related modular calcium-binding protein 1 gene, SMOC1, cause waardenburg anophthalmia syndrome.
Mutations in the SPARC-related modular calcium-binding protein 1 gene, SMOC1, cause waardenburg anophthalmia syndrome.
American journal of human genetics (2011-01-05)
Hana Abouzeid, Gaëlle Boisset, Tatiana Favez, Mohamed Youssef, Iman Marzouk, Nihal Shakankiry, Nader Bayoumi, Patrick Descombes, Céline Agosti, Francis L Munier, Daniel F Schorderet
ABSTRACT
Waardenburg anophthalmia syndrome, also known as microphthalmia with limb anomalies, ophthalmoacromelic syndrome, and anophthalmia-syndactyly, is a rare autosomal-recessive developmental disorder that has been mapped to 10p11.23. Here we show that this disease is heterogeneous by reporting on a consanguineous family, not linked to the 10p11.23 locus, whose two affected children have a homozygous mutation in SMOC1. Knockdown experiments of the zebrafish smoc1 revealed that smoc1 is important in eye development and that it is expressed in many organs, including brain and somites.