Merck
  • Reductive and brominative termination of alkenol cyclization in aerobic cobalt-catalyzed reactions.
Reductive and brominative termination of alkenol cyclization in aerobic cobalt-catalyzed reactions.
Journal of the American Chemical Society (2009-08-22)
Dominik Schuch, Patrick Fries, Maike Dönges, Bárbara Menéndez Pérez, Jens Hartung
ABSTRACT
Tetrahydrofur-2-ylmethyl radicals were stereoselectively generated from substituted pent-4-en-1-ols in aerobic cobalt(II)-catalyzed oxidations. Intermediates were trapped with cyclohexa-1,4-diene, gamma-terpinene, BrCCl(3), diethyl dibromomalonate, or electron-deficient olefins such as acrylonitrile or dimethyl fumarate to afford functionalized tetrahydrofurans in synthetically useful yields.
Sigma-Aldrich
Diethyl dibromomalonate, 97%