Merck
  • Diagnosis of Burkitt lymphoma using an algorithmic approach--applicable in both resource-poor and resource-rich countries.
Diagnosis of Burkitt lymphoma using an algorithmic approach--applicable in both resource-poor and resource-rich countries.
British journal of haematology (2011-07-02)
Kikkeri N Naresh, Hazem A H Ibrahim, Stefano Lazzi, Patricia Rince, Monica Onorati, Maria R Ambrosio, Chrystèle Bilhou-Nabera, Furrat Amen, Alistair Reid, Michael Mawanda, Valeria Calbi, Martin Ogwang, Emily Rogena, Bessie Byakika, Shahin Sayed, Emma Moshi, Amos Mwakigonja, Martine Raphael, Ian Magrath, Lorenzo Leoncini
ABSTRACT
Distinguishing Burkitt lymphoma (BL) from B cell lymphoma, unclassifiable with features intermediate between diffuse large B-cell lymphoma (DLBCL) and BL (DLBCL/BL), and DLBCL is challenging. We propose an immunohistochemistry and fluorescent in situ hybridization (FISH) based scoring system that is employed in three phases - Phase 1 (morphology with CD10 and BCL2 immunostains), Phase 2 (CD38, CD44 and Ki-67 immunostains) and Phase 3 (FISH on paraffin sections for MYC, BCL2, BCL6 and immunoglobulin family genes). The system was evaluated on 252 aggressive B-cell lymphomas from Europe and from sub-Saharan Africa. Using the algorithm, we determined a specific diagnosis of BL or not-BL in 82%, 92% and 95% cases at Phases 1, 2 and 3, respectively. In 3·4% cases, the algorithm was not completely applicable due to technical reasons. Overall, this approach led to a specific diagnosis of BL in 122 cases and to a specific diagnosis of either DLBCL or DLBCL/BL in 94% of cases that were not diagnosed as BL. We also evaluated the scoring system on 27 cases of BL confirmed on gene expression/microRNA expression profiling. Phase 1 of our scoring system led to a diagnosis of BL in 100% of these cases.