Merck
  • Adsorptive stripping voltammetric determination of 4-aminophenol at a single-wall carbon nanotubes film coated electrode.
Adsorptive stripping voltammetric determination of 4-aminophenol at a single-wall carbon nanotubes film coated electrode.
Talanta (2008-10-31)
Wensheng Huang, Weibing Hu, Jinchun Song
ABSTRACT
A single-wall carbon nanotubes (SWNT)-Nafion film coated glassy carbon electrode (GCE) was described for the determination of 4-aminophenol. In pH 3.0 sodium citrate-HCl buffer, the oxidation peak current of 4-aminophenol increases greatly at the SWNT-Nafion film coated GCE in contrast to that at both bare GCE and Nafion-film coated GCE. Moreover, the oxidation peak potential shifts to more negative potential. All the experimental parameters were optimized for the determination of 4-aminophenol. The oxidation peak current is proportional to the concentration of 4-aminophenol over the range from 5x10(-9) to 2x10(-6) mol l(-1). The detection limit is 8x10(-10) mol l(-1) at 4 min of accumulation. Using the proposed method, 4-aminophenol in water samples was determined.
Sigma-Aldrich
4-Aminophenol, ≥98%