Merck
  • Flow cytometric determination of genome size in European sunbleak Leucaspius delineatus (Heckel, 1843).
Flow cytometric determination of genome size in European sunbleak Leucaspius delineatus (Heckel, 1843).
Fish physiology and biochemistry (2011-05-27)
Marta Filipiak, Grzegorz Tylko, Wincenty Kilarski
ABSTRACT
The aim of this study was to compare DNA content in hepatocyte and erythrocyte nuclei of the European sunbleak, Leucaspius delineatus, in relation to nuclear and cell size by means of flow cytometry and fluorescence microscopy. The DNA standards, chicken and rainbow trout erythrocytes, were prepared in parallel with both cell types, with initial separation of liver cells in pepsin solution followed by cell filtering. Standards and investigated cells were stained with a mixture of propidium iodide, citric acid, and Nonidet P40 in the presence of RNAse, and fluorescence of at least 50,000 nuclei was analyzed by flow cytometry. Average cell size was determined by flow cytometry, using fresh cell suspension in relation to latex beads of known diameter. The size of nuclei was examined on the basis of digital micrographs obtained by fluorescence microscopy after nuclei staining with DAPI. The sunbleak's erythrocyte nuclei contain 2.25 ± 0.06 pg of DNA, whereas the hepatocyte nuclei contain 2.46 ± 0.06 pg of DNA. This difference in DNA content was determined spectroscopically using isolated DNA from the two cell types. The modal diameters of the erythrocytes and hepatocytes were estimated to be 5.1 ± 0.2 and 22.3 ± 5.0 μm, respectively, and the corresponding modal dimensions of their nuclei (measured as surface area) were 15.2 and 21.4 μm(2), respectively. The nucleoplasmic index, as calculated from diameters estimated from surface area of nuclear profiles, was 2.51 for the erythrocytes compared with 0.08 for hepatocytes.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
SAFC
Hanks′ Balanced Salts, Modified, without calcium chloride, magnesium sulfate, phenol red and sodium bicarbonate, powder, suitable for cell culture