Merck
  • An environmentally benign synthesis of cis-2,6-disubstituted tetrahydropyrans via indium-mediated tandem allylation/Prins cyclization reaction.
An environmentally benign synthesis of cis-2,6-disubstituted tetrahydropyrans via indium-mediated tandem allylation/Prins cyclization reaction.
The Journal of organic chemistry (2007-12-22)
Minh Pham, Amir Allatabakhsh, Thomas G Minehan
ABSTRACT
In the presence of indium metal, 3-iodo-2-[(trimethylsilyl)methyl]propene (1) reacts with sequentially added aldehydes to provide cis-2,6-disubstituted tetrahydropyrans in good yields. Evidence suggests that InI, formed upon aldehyde (R1CHO) allylation in aqueous media, acts as a promoter for the silyl-Prins reaction with the second equivalent of added aldehyde (R2CHO). The preparation of cyclohexenyl-fused pyrans via this one-pot, three-component coupling process is presented, as is a short formal synthesis of (+/-)-centrolobine.
Sigma-Aldrich
trans-3-(Trimethylsilyl)allyl alcohol, 96%