Merck
  • GTPase activating specificity of RGS12 and binding specificity of an alternatively spliced PDZ (PSD-95/Dlg/ZO-1) domain.
GTPase activating specificity of RGS12 and binding specificity of an alternatively spliced PDZ (PSD-95/Dlg/ZO-1) domain.
The Journal of biological chemistry (1998-07-04)
B E Snow, R A Hall, A M Krumins, G M Brothers, D Bouchard, C A Brothers, S Chung, J Mangion, A G Gilman, R J Lefkowitz, D P Siderovski
ABSTRACT
Regulator of G-protein signaling (RGS) proteins increase the intrinsic guanosine triphosphatase (GTPase) activity of G-protein alpha subunits in vitro, but how specific G-protein-coupled receptor systems are targeted for down-regulation by RGS proteins remains uncharacterized. Here, we describe the GTPase specificity of RGS12 and identify four alternatively spliced forms of human RGS12 mRNA. Two RGS12 isoforms of 6.3 and 5.7 kilobases (kb), encoding both an N-terminal PDZ (PSD-95/Dlg/ZO-1) domain and the RGS domain, are expressed in most tissues, with highest levels observed in testis, ovary, spleen, cerebellum, and caudate nucleus. The 5.7-kb isoform has an alternative 3' end encoding a putative C-terminal PDZ domain docking site. Two smaller isoforms, of 3.1 and 3.7 kb, which lack the PDZ domain and encode the RGS domain with and without the alternative 3' end, respectively, are most abundantly expressed in brain, kidney, thymus, and prostate. In vitro biochemical assays indicate that RGS12 is a GTPase-activating protein for Gi class alpha subunits. Biochemical and interaction trap experiments suggest that the RGS12 N terminus acts as a classical PDZ domain, binding selectively to C-terminal (A/S)-T-X-(L/V) motifs as found within both the interleukin-8 receptor B (CXCR2) and the alternative 3' exon form of RGS12. The presence of an alternatively spliced PDZ domain within RGS12 suggests a mechanism by which RGS proteins may target specific G-protein-coupled receptor systems for desensitization.