Merck
  • Effects of chromophore orientation and molecule conformation on surface-enhanced Raman scattering studied with alkanoic acids and colloidal silver nanoparticles.
Effects of chromophore orientation and molecule conformation on surface-enhanced Raman scattering studied with alkanoic acids and colloidal silver nanoparticles.
The Journal of chemical physics (2006-12-28)
Leo Seballos, Tammy Y Olson, Jin Z Zhang
ABSTRACT
Experimental studies have been carried out to gain a better understanding of the effects of chromophore orientation and molecular conformation on surface-enhanced Raman scattering (SERS) based on metal nanostructures. A series of alkanoic acids that contain a phenyl ring separated by methylene groups from the carboxylic acid, including phenylacetic acid, 3-phenylpropionic acid, 4-phenylbutyric acid, 5-phenylvaleric acid, and 6-phenylhexanoic acid, was investigated as model molecules with colloidal silver nanoparticles as SERS substrates. As the number of methylene groups increases, the molecules display an interesting zigzag intensity pattern of the phenyl ring bending mode around 1000 cm(-1) as well as a trend of appearance and disappearance of either the degenerate ring breathing mode or C[Double Bond]O vibrational mode near 1585 and 1630 cm(-1), respectively. Molecules containing an odd number of methylene units display a higher ring bending intensity and degenerate ring breathing mode and are suggested to have a trans conformation on the particle surface. Molecules with an even number of methylene units show a C[Double Bond]O vibrational mode and weaker ring bending in their SERS spectra and are suggested to have a gauche conformation on the silver nanoparticle surface. The different conformation is attributed to the varying interactions of the carboxylic group or the phenyl ring pi electrons with the silver surface. The SERS intensity was found to change little as the length between the phenyl ring and the carboxylic group was increased by adding CH(2) spacers. This is possibly because the effective distance between the phenyl ring and the silver surface does not change much with increasing number of CH(2) spacers as a result of changes in molecular conformation and variations in the phenyl ring orientation with CH(2) addition. The insight gained from this study is important for understanding SERS of complex molecules for which chromophore orientation and molecular conformation must be taken into careful consideration.
Sigma-Aldrich
6-Phenylhexanoic acid, 98%