Merck
Haemodynamics and tissue specificity with isradipine.
Acta anaesthesiologica Scandinavica. Supplementum (1993-01-01)
J B Leslie
ABSTRACT
The significant effects of isradipine in producing arterial vasodilation and mild negative chronotropic effects without significant negative inotropic effects suggests that this agent should provide excellent antihypertensive efficacy in the treatment of perioperative hypertension. Isradipine may prove to be a safe antihypertensive treatment in patients with impaired ventricular function (cardiac failure), impaired myocardial perfusion (ischaemia), and in cases of selected conduction abnormalities or arrhythmias. The demonstration that its effects are limited to vascular resistance rather than vascular capacitance is an important distinguishing feature of isradipine compared with other antihypertensive agents. Finally, the potential application of this dihydropyridine calcium antagonist for cytoprotection and its effects on atherosclerosis remain exciting therapeutic prospects.
Sigma-Aldrich
Isradipine, ≥98% (HPLC), solid
SKU
Pack Size
Availability
Price
Quantity
Isradipine, European Pharmacopoeia (EP) Reference Standard