- Miglitol: assessment of its role in the treatment of patients with diabetes mellitus.
Miglitol: assessment of its role in the treatment of patients with diabetes mellitus.
To evaluate miglitol, a new oral alpha-glucosidase inhibitor, and discuss its pharmacology, therapeutics, pharmacokinetics, dosing guidelines, adverse effects, drug interactions, and clinical efficacy. A MEDLINE English-language only database search using the keywords miglitol, glyset, and Bay m 1099 (1985 to December 1999), was completed to identify relevant articles including reviews, recent studies, and abstracts; American Diabetes Association 1999 Annual Meeting abstracts; Pharmacia & Upjohn data on file and product information. The clinical trials that were selected to be reviewed in detail were randomized, double-blind studies with at least 100 patients in the intention-to-treat group. All articles and abstracts were reviewed along with the product labeling from Pharmacia & Upjohn. Miglitol is an alpha-glucosidase inhibitor that exerts its effect through the delayed absorption of complex carbohydrates in the small intestine, resulting in a decrease in postprandial glucose concentrations that are directly correlated with the dietary carbohydrate content. Both small, short-term trials and large, clinical trials show a decrease in postprandial glucose concentrations and a modest decrease in glycosylated hemoglobin of approximately 0.5-1.0% as a result of miglitol's action. The adverse effects of miglitol are mild and transitory and include flatulence, diarrhea, and abdominal pain. The incidence of gastrointestinal problems may be reduced with a small initial dose, which is slowly titrated as tolerated. Miglitol is an effective and safe treatment option in patients with type 2 diabetes mellitus who are inadequately controlled with diet or oral sulfonylurea therapy. Miglitol is a good choice of therapy in Hispanic, African-American, and elderly patients, or any patients in whom hypoglycemia, weight gain, or lactic acidosis are risks. No published studies comparing miglitol with acarbose have been published, but there appears to be no major clinical or financial advantages to using one agent over the other.