Merck
Total synthesis of (-)-incrustoporin.
Journal of Asian natural products research (2001-03-20)
Q Yu, J F Liu, Y L Wu
ABSTRACT
(-)-Incrustoporin (1) has been synthesized using aldol condensation of ethyl p-tolyl-acetate (2) and (2R)-benzoyloxy-butanal (3), followed by acid-catalyzed deprotection of the benzoyl group, lactone ring-closure, and elimination of the beta-OH in a one-pot manner. The aldehyde 3 was prepared from the commercially available D-mannitol by a two-directional strategy.
Sigma-Aldrich
p-Tolyl acetate, 99%