Merck
  • Self-immobilized catalysts for ethylene polymerization: neutral, single-component salicylaldiminato phenyl nickel(II) complexes bearing allyl substituents.
Self-immobilized catalysts for ethylene polymerization: neutral, single-component salicylaldiminato phenyl nickel(II) complexes bearing allyl substituents.
Chemical communications (Cambridge, England) (2002-07-18)
Dao Zhang, Guo-Xin Jin, Ninghai Hu
ABSTRACT
A new family of self-immobilized ethylene polymerization catalysts, derived from neutral, single-component salicylaldiminato phenyl nickel complexes, is described.