Merck
  • Investigation on physicochemical and biological differences of cefpodoxime proxetil enantiomers.
Investigation on physicochemical and biological differences of cefpodoxime proxetil enantiomers.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2006-06-27)
Vasu Kumar Kakumanu, Vinod Arora, Arvind K Bansal
ABSTRACT
Cefpodoxime proxetil (CP) is a prodrug of cefpodoxime acid (CA), and is supplied as racemic mixture of R- and S-enantiomers. CP has only 50% absolute bioavailability, and the reasons responsible for low bioavailability remain poorly understood. The present work ascertains physicochemical and biological properties of individual isomers of CP and explores their capacity to optimize delivery of CP. Both isomers showed similar pH stability behavior, but R-isomer was more susceptible to enzymatic metabolism compared to S-isomer, when incubated with enzymes collected from various segments of GIT. Based on the in vitro and in vivo results, use of S-isomer for development of a dosage form such as gastro-retentive dosage form can improve oral bioavailability of CP.
Product Description
Cefpodoxime proxetil for impurity H identification, European Pharmacopoeia (EP) Reference Standard
Cefpodoxime proxetil for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Cefpodoxime proxetil, United States Pharmacopeia (USP) Reference Standard
Cefpodoxime proxetil, European Pharmacopoeia (EP) Reference Standard