Merck
  • Facile synthesis of phosphonamidate- and phosphonate-linked phosphonopeptides.
Facile synthesis of phosphonamidate- and phosphonate-linked phosphonopeptides.
Journal of peptide science : an official publication of the European Peptide Society (2005-10-26)
Nanyan Fu, Qihan Zhang, Lifang Duan, Jiaxi Xu
ABSTRACT
A direct method for the preparation of phosphonamidate- and phosphonate-linked phosphonopeptides has been developed. Using this method, both phosphonopeptides were prepared in acceptable yields directly from simple and commercially available chemicals in one-pot reactions of benzyl carbamate, aldehydes, and methyl dichlorophosphite, followed by aminolysis with amino acid esters or alcoholysis with hydroxy esters.
Sigma-Aldrich
Methyl dichlorophosphite, technical grade
Pricing and availability is not currently available.