Merck
  • Immunization of Saimiri sciureus monkeys with Plasmodium falciparum merozoite surface protein-3 and glutamate-rich protein suggests that protection is related to antibody levels.
Immunization of Saimiri sciureus monkeys with Plasmodium falciparum merozoite surface protein-3 and glutamate-rich protein suggests that protection is related to antibody levels.
Scandinavian journal of immunology (2004-03-31)
L J M Carvalho, S G Oliveira, M Theisen, F A Alves, M C R Andrade, G M Zanini, M C O Brígido, C Oeuvray, M M Póvoa, J A P C Muniz, P Druilhe, C T Daniel-Ribeiro
ABSTRACT
The immunogenicity and protective efficacy of various antigen-adjuvant formulations derived either from the merozoite-surface protein-3 (MSP-3) or the glutamate-rich protein (GLURP) of Plasmodium falciparum were evaluated in Saimiri sciureus monkeys. These proteins were selected for immunogenicity studies based primarily on their capacity of inducing an antibody-dependent cellular inhibition effect on parasite growth. Some of the S. sciureus monkeys immunized with MSP-3(212-380)-AS02 or GLURP(27-500)-alum were able to fully or partially control parasitaemia upon an experimental P. falciparum [Falciparum Uganda Palo Alto (FUP-SP) strain] blood-stage infection, and this protection was related to the prechallenge antibody titres induced. The data are indicative that MSP-3 and GLURP can induce protective immunity against an experimental P. falciparum infection using adjuvants that are acceptable for human use and this should trigger further studies with those new antigens.