Merck
  • Volatile sulfur compounds in mouth air from clinically healthy subjects and patients with periodontal disease.
Volatile sulfur compounds in mouth air from clinically healthy subjects and patients with periodontal disease.
Journal of periodontal research (1992-07-01)
K Yaegaki, K Sanada
ABSTRACT
Volatile sulfur compounds (VSC) in mouth air were estimated by gas chromatography. The amount of VSC and the methyl mercaptan/hydrogen sulfide ratio were significantly increased in patients with periodontal disease. These two parameters also increased in proportion to the bleeding index and probing depth. A study was also done on the effect of removal of tongue coating on VSC concentrations in mouth air from patients with periodontal involvement. VSC and the methyl mercaptan/hydrogen sulfide ratio were reduced to 49% and 35%, respectively, by removal of the tongue coating. The average amount of tongue coating removed from patients with periodontal disease was significantly higher than from controls (90.1 mg vs. 14.6 mg, p less than 0.01). Estimated production of VSC from tongue coating was 4 times higher than the control value, and the methyl mercaptan/hydrogen sulfide ratio was also markedly increased. However, a saliva putrefaction study suggested that saliva does not contribute to the elevated ratio of methyl mercaptan in mouth air. These results strongly suggest that, in addition to periodontal pockets, tongue coating has an important role in VSC production, in particular leading to an elevated concentration of methyl mercaptan, which is more pathogenic than hydrogen sulfide.
Sigma-Aldrich
Methanethiol, ≥98.0%