Merck
  • Progesterone metabolism in human fibroblasts is independent of P-glycoprotein levels and Niemann-Pick type C disease.
Progesterone metabolism in human fibroblasts is independent of P-glycoprotein levels and Niemann-Pick type C disease.
The Journal of steroid biochemistry and molecular biology (2000-01-06)
J Zhang, L J Ming, J Sjövall, H W Cook, N D Ridgway, D M Byers
ABSTRACT
Progesterone inhibits intracellular transport of lysosomal cholesterol in cultured cells, and thus at least in part mimics the biochemical phenotype of Niemann-Pick type C disease (NPC) in human fibroblasts. The goal of this study was to determine whether metabolism of progesterone to other steroids is affected by the NPC mutation or by P-glycoprotein (a known progesterone target). We found that human fibroblasts metabolize progesterone in three steps: rapid conversion to 5alpha-pregnane-3,20-dione, which is then reduced to 5alpha-pregnane-3beta(alpha)-ol-20-one with subsequent 6alpha-hydroxylation. The pattern and rates of progesterone metabolism were not significantly different in a variety of fibroblasts from normal individuals, NPC patients, and obligate heterozygotes. Inhibition of steroid 5alpha-reductase with finasteride completely blocked metabolism of progesterone but had no effect on inhibition of LDL-stimulated cholesterol esterification (IC50 = 10 microM). Progesterone also partially inhibited 25-hydroxycholesterol-induced cholesterol esterification, with similar dose-dependence in normal and NPC fibroblasts. P-glycoprotein levels varied significantly among the various fibroblasts tested, but no correlation with NPC phenotype or rate of progesterone metabolism was noted, and P-glycoprotein inhibitors did not affect conversion of progesterone to products. These results indicate that metabolism of progesterone in human fibroblasts is largely independent of its ability to interfere with cholesterol traffic and P-glycoprotein function.
Sigma-Aldrich
5α-Pregnane-3,20-dione (allo), progesterone metabolite