Merck
  • Oxidation of phenanthrene by a fungal laccase in the presence of 1-hydroxybenzotriazole and unsaturated lipids.
Oxidation of phenanthrene by a fungal laccase in the presence of 1-hydroxybenzotriazole and unsaturated lipids.
Biochemical and biophysical research communications (1998-03-26)
S Böhmer, K Messner, E Srebotnik
ABSTRACT
Phenanthrene, a polycyclic aromatic hydrocarbon, was efficiently oxidized by laccase in the presence of both 1-hydroxybenzotriazole and unsaturated lipids. 73% of initially added phenanthrene was degraded within 182 hours to give phenanthrene-9,10-quinone and 2,2'-diphenic acid as the major products. The system was also able to peroxidize linoleic acid to its corresponding hydroperoxides suggesting the involvement of lipid peroxidation in laccase catalyzed phenanthrene oxidation. Lipid peroxidation by laccase required 1-hydroxybenzotriazole and did not depend on Mn2+ and H2O2 suggesting that the chemical reactions involved differ from those previously reported for manganese peroxidase.
Sigma-Aldrich
Diphenic acid, 97%