Merck
Computational design of a substrate specificity mutant of a protein.
Proteins (1996-12-01)
N Honda, Y Komeiji, M Uebayasi, I Yamato
ABSTRACT
The wild-type trp repressor of E. coli bound 5-methoxytryptophan, a Trp analogue, less tightly than Trp. A mutant repressor (Val58-->Ala) that should bind 5-methoxytryptophan preferentially to Trp was computationally designed by free-energy calculations accompanied by free-energy decomposition. The designed mutant was demonstrated by experiments to bind 5-methoxytryptophan more tightly than Trp, consistent with the computational prediction. This success indicates the usefulness of free energy decomposition in protein design.
Sigma-Aldrich
5-Methoxy-DL-tryptophan