Merck
  • Direct enantiomeric resolution of mephenytoin and its N-demethylated metabolite in plasma and blood using chiral capillary gas chromatography.
Direct enantiomeric resolution of mephenytoin and its N-demethylated metabolite in plasma and blood using chiral capillary gas chromatography.
Journal of chromatography (1984-04-13)
P J Wedlund, B J Sweetman, C B McAllister, R A Branch, G R Wilkinson
ABSTRACT
A gas chromatographic method was developed for the determination of the R- and S- enantiomers of the anticonvulsant, mephenytoin, and its N-demethylated metabolite, 5-phenyl-5- ethylhydantoin ( PEH ), in plasma and blood. Direct enantiomeric separation of mephenytoin and its internal standard was obtained using a chiral capillary column ( Chirasil -Val) followed by nitrogen specific detection. However, resolution of the enantiomers of PEH and its internal standard required propylation at the 3-position of the hydantoin ring prior to analysis. Similar linear and reproducible standard curves were obtained from both plasma and blood over the concentration range 50 ng/ml to 5 micrograms/ml, and above 100 ng/ml the reproducibility was less than 8% (coefficient of variation). Pronounced stereoselective differences in the plasma concentration--time curves for both mephenytoin and PEH were observed in a normal subject who received a single oral dose of 300 mg racemic mephenytoin. The peak plasma level of S-mephenytoin was only one-fifth that of the R-enantiomer and its elimination half-life was less than 3 h compared to over 70 h for R-mephenytoin. Similarly, S- PEH levels were barely detectable whereas concentrations of R-metabolite steadily increased over 4-6 days before slowly declining.
Sigma-Aldrich
(R)-(−)-Nirvanol
Sigma-Aldrich
(S)-(+)-Nirvanol