- Encapsulation of triphenyltin hydroxide.
Encapsulation of triphenyltin hydroxide.
Journal of microencapsulation (1986-01-01)
A Markus, S Felix, Z Pelah
PMID3508175
ABSTRACT
Triphenyltin hydroxide was encapsulated to prolong its fungicidal activity and to decrease its phytotoxicity in peanut fields. The envelopes found to be adequate for this purpose were polyureas and ethyl cellulose, as was demonstrated by biological tests.
MATERIALS